3,5-Dimethylanisole 98%

Supplier: Ambeed
Warning

A324494-100G A324494-1G A324494-25G A324494-5G
77277-754EA 72 USD
77277-754 77277-756 77277-758 77277-760
3,5-Dimethylanisole 98%
3,5-Dimethylanisole

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00008398
CAS Number: 874-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR