7-((S)-7-Amino-5-azaspiro[2.4]heptan-5-yl)-8-chloro-6-fluoro-1-((1R,2S)-2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid Sesquihydrate 99%
Supplier: Ambeed
|
Warning
|
A324321-10MG
A324321-1MG
A324321-50MG
A324321-5MG
A324321-250MG
A324321-100MG
77239-648EA
36
USD
77239-648
77239-650
77239-652
77239-654
77746-631
77746-630
7-((S)-7-Amino-5-azaspiro[2.4]heptan-5-yl)-8-chloro-6-fluoro-1-((1R,2S)-2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid Sesquihydrate 99%
7-((S)-7-Amino-5-azaspiro[2.4]heptan-5-yl)-8-chloro-6-fluoro-1-((1R,2S)-2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid Sesquihydrate
Precaución: for Research Use Only
Formula:
C₃₈H₄₂Cl₂F₄N₆O₉ |
MDL Number:
MFCD11046360 CAS Number: 163253-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...