4-(Hydroxymethyl)indole 97%

Supplier: Ambeed
Warning

Synonyms: (1H-Indol-4-yl)methanol

A324244-100MG A324244-250MG
77277-750EA 14.42 USD
77277-750 77277-752
4-(Hydroxymethyl)indole 97%
4-(Hydroxymethyl)indole

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.1739 g/mol
MDL Number: MFCD01632220
CAS Number: 1074-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR