1H-Pyrrolo[3,2-c]pyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A324087-100MG A324087-1G A324087-250MG
77277-744EA 36.05 USD
77277-744 77277-746 77277-748
1H-Pyrrolo[3,2-c]pyridine-3-carboxylic acid 97%
1H-Pyrrolo[3,2-c]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD08690129
CAS Number: 119248-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR