4-Chloro-2-methyl-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine 97%

Supplier: Ambeed
Danger

A323844-1G A323844-250MG
77239-642EA 153 USD
77239-642 77239-644
4-Chloro-2-methyl-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine 97%
4-Chloro-2-methyl-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S
MDL Number: MFCD00572401
CAS Number: 300816-24-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR