4-(Benzyloxy)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxybenzeneboronic acid pinacol ester, 2-(4-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A323812-1G A323812-250MG A323812-5G
77290-332EA 6.18 USD
77290-332 77290-334 77290-336
4-(Benzyloxy)phenylboronic acid pinacol ester 97%
4-(Benzyloxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₃BO₃
MW: 310.19 g/mol
MDL Number: MFCD04974107
CAS Number: 754226-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR