3-Aminopyridin-2-ol 98%

Supplier: Ambeed
Warning

A323675-100G A323675-5G A323675-25G
77290-328EA 501 USD
77290-328 77290-330 77748-635
3-Aminopyridin-2-ol 98%
3-Aminopyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MDL Number: MFCD03411556
CAS Number: 59315-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR