2,3-Difluoro-6-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A323435-1G A323435-250MG A323435-5G
77239-630EA 10 USD
77239-630 77239-632 77239-634
2,3-Difluoro-6-methoxybenzaldehyde 97%
2,3-Difluoro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD02093968
CAS Number: 187543-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR