3-Bromo-6-chloro-2-methyl-5-nitropyridine 97%

Supplier: Ambeed
Warning

A323170-10G A323170-1G A323170-250MG A323170-5G
77263-468EA 77 USD
77263-468 77263-470 77263-472 77263-474
3-Bromo-6-chloro-2-methyl-5-nitropyridine 97%
3-Bromo-6-chloro-2-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O₂
MDL Number: MFCD09839274
CAS Number: 186413-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR