1,1,2,2-Tetrakis(4-(pyridin-4-yl)phenyl)ethene 97%

Supplier: Ambeed
Warning

A323135-100MG A323135-1G A323135-250MG A323135-5G
77263-460EA 54.59 USD
77263-460 77263-462 77263-464 77263-466
1,1,2,2-Tetrakis(4-(pyridin-4-yl)phenyl)ethene 97%
1,1,2,2-Tetrakis(4-(pyridin-4-yl)phenyl)ethene

Precaución: for Research Use Only

Formula: C₄₆H₃₂N₄
MW: 640.77 g/mol
MDL Number: MFCD31692919
CAS Number: 1227195-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR