3-Chloro-4-fluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-fluorobenzyl Alcohol, (3-Chloro-4-fluorophenyl)methanol

A322980-1G A322980-25G
77277-734EA 8.24 USD
77277-734 77277-736
3-Chloro-4-fluorobenzyl alcohol 98%
3-Chloro-4-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD01631550
CAS Number: 161446-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR