1-Chloro-2,4,6-trifluorobenzene 95%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-1,3,5-trifluorobenzene, 2,4,6-Trifluorochlorobenzene

A322836-1G A322836-25G
77239-624EA 20.6 USD
77239-624 77746-628
1-Chloro-2,4,6-trifluorobenzene 95%
1-Chloro-2,4,6-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₃
MW: 166.528 g/mol
MDL Number: MFCD00012239
CAS Number: 2106-40-3
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR