2-Methoxyethyl acetoacetate 96%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxyethyl 3-oxobutanoate

A322512-100G A322512-25G A322512-500G A322512-5G A322512-1000G
77263-440EA 39 USD
77263-440 77263-442 77263-444 77263-446 77616-422
2-Methoxyethyl acetoacetate 96%
2-Methoxyethyl acetoacetate

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MDL Number: MFCD00009648
CAS Number: 22502-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR