2'-Deoxycytidine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
A322509-1G
A322509-25G
A322509-5G
77251-170EA
20.6
USD
77251-170
77251-172
77251-174
2'-Deoxycytidine hydrochloride 98%
2'-Deoxycytidine hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₄ClN₃O₄ MW: 263.68 g/mol |
MDL Number:
MFCD00012840 CAS Number: 3992-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...