Tris[4,4'-di-tert-butyl-(2,2')-bipyridine]ruthenium(III) dihexafluorophosphate 97%
Supplier: Ambeed
|
Warning
|
A322445-100MG
A322445-1G
A322445-250MG
A322445-50MG
77251-162EA
40
USD
77251-162
77251-164
77251-166
77251-168
Tris[4,4'-di-tert-butyl-(2,2')-bipyridine]ruthenium(III) dihexafluorophosphate 97%
Tris[4,4'-di-tert-butyl-(2,2')-bipyridine]ruthenium(III) dihexafluorophosphate
Precaución: for Research Use Only
Formula:
C₅₄H₇₂F₁₂N₆P₂Ru MW: 1196.18 g/mol |
MDL Number:
MFCD12022556 CAS Number: 75777-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...