2,6-Dimethylnaphthalene 98%

Supplier: Ambeed
Danger

A322385-1G A322385-250MG A322385-5G
77251-158EA 45 USD
77251-158 77251-160 77550-898
2,6-Dimethylnaphthalene 98%
2,6-Dimethylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 169.8
Melting Pt: -44.7
MDL Number: MFCD00004120
CAS Number: 581-42-0
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR