2-Methyl-3-furoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2-Methylfuran-3-carboxylic acid

A322182-1G A322182-25G A322182-5G
77277-714EA 11.33 USD
77277-714 77277-716 77277-718
2-Methyl-3-furoic acid 97%
2-Methyl-3-furoic acid

Precaución: for Research Use Only

Formula: C₆H₆O₃
MW: 126.11 g/mol
MDL Number: MFCD00092314
CAS Number: 6947-94-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR