(R)-tert-Butyl (1-(4-bromophenyl)-2-hydroxyethyl)carbamate 97%

Supplier: Ambeed
Warning

A322075-100MG A322075-1G A322075-250MG A322075-5G
77263-422EA 26.78 USD
77263-422 77263-424 77263-426 77263-428
(R)-tert-Butyl (1-(4-bromophenyl)-2-hydroxyethyl)carbamate 97%
(R)-tert-Butyl (1-(4-bromophenyl)-2-hydroxyethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₃
MW: 316.19 g/mol
MDL Number: MFCD28148212
CAS Number: 849178-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR