4-(4-Fluorophenoxy)benzoic acid 95%

Supplier: Ambeed
Warning

A321760-1G A321760-250MG A321760-5G A321760-10G
77290-300EA 10 USD
77290-300 77290-302 77290-304 77748-631
4-(4-Fluorophenoxy)benzoic acid 95%
4-(4-Fluorophenoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₉FO₃
MDL Number: MFCD01631892
CAS Number: 129623-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR