tert-Butyl 1-hydroxy-3,6,9,12,15,18-hexaoxahenicosan-21-oate 98%

Supplier: Ambeed
Warning

Synonyms: Hydroxy-PEG6-Boc

A321620-1G A321620-250MG A321620-5G A321620-25G
77263-418EA 150.38 USD
77263-418 77263-420 77568-646 77748-630
tert-Butyl 1-hydroxy-3,6,9,12,15,18-hexaoxahenicosan-21-oate 98%
tert-Butyl 1-hydroxy-3,6,9,12,15,18-hexaoxahenicosan-21-oate

Precaución: For research use only.

Formula: C₁₉H₃₈O₉
MW: 410.49 g/mol
MDL Number: MFCD13185017
CAS Number: 361189-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR