(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate 97%

Supplier: Ambeed
Warning

Synonyms: 2',3',5'-Tri-O-acetylinosine

A321426-1G A321426-250MG A321426-5G A321426-25G A321426-10G
77277-708EA 10 USD
77277-708 77277-710 77277-712 77748-629 77748-628
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate 97%
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diyl diacetate

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₄O₈
MDL Number: MFCD00038617
CAS Number: 3181-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR