tert-Butyl 4-(hydroxymethyl)benzoate 98%

Supplier: Ambeed
Warning

A321260-100MG A321260-250MG A321260-5G
77290-294EA 76.22 USD
77290-294 77290-296 77290-298
tert-Butyl 4-(hydroxymethyl)benzoate 98%
tert-Butyl 4-(hydroxymethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
MDL Number: MFCD04973449
CAS Number: 143726-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR