(R)-3,3'-Bis(2-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate ≥98%, ee 99%

Supplier: Ambeed
Warning

A321157-1G A321157-250MG
77251-146EA 501 USD
77251-146 77251-148
(R)-3,3'-Bis(2-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate ≥98%, ee 99%
(R)-3,3'-Bis(2-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate

Precaución: for Research Use Only

Formula: C₄₀H₂₅O₄P
MDL Number: MFCD17676198
CAS Number: 791616-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR