1-Methylisatin 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylindoline-2,3-dione, 1-Methyl-2,3-dihydroindole-2,3-dione, N-Methylindoline-2,3-dione, N-Methylisatin, 1-Methyl-1H-indole-2,3-dione, N-Methyl-1H-indole-2,3-dione, N-Methyl-2,3-dihydroindole-2,3-dione

A321065-100G A321065-10G A321065-1G A321065-250MG A321065-25G A321065-5G
77263-406EA 511.91 USD
77263-406 77263-408 77263-410 77263-412 77263-414 77263-416
1-Methylisatin 98%
1-Methylisatin

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00005812
CAS Number: 2058-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR