6-Methoxy-2-methylquinolin-5-amine 97%
Supplier: Ambeed
|
Warning
|
A321058-100MG
A321058-1G
A321058-250MG
77239-586EA
164.8
USD
77239-586
77239-588
77239-590
6-Methoxy-2-methylquinolin-5-amine 97%
6-Methoxy-2-methylquinolin-5-amine
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O MW: 188.22 g/mol |
MDL Number:
MFCD08690735 CAS Number: 114656-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...