2-(4-Methoxybenzoyl)thiophene 98%

Supplier: Ambeed
Warning

Synonyms: (4-Methoxyphenyl)(thiophen-2-yl)methanone, 4-Methoxyphenyl 2-Thienyl Ketone

A320704-1G A320704-5G
77239-578EA 16.48 USD
77239-578 77616-416
2-(4-Methoxybenzoyl)thiophene 98%
2-(4-Methoxybenzoyl)thiophene

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂S
MW: 218.27 g/mol
MDL Number: MFCD00005440
CAS Number: 4160-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR