(S)-2-(2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
A320529-1G
A320529-25G
A320529-5G
77251-122EA
39
USD
77251-122
77251-124
77251-126
(S)-2-(2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl)acetic acid 95%
(S)-2-(2,2-Dimethyl-5-oxo-1,3-dioxolan-4-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀O₅ |
MDL Number:
MFCD00211273 CAS Number: 73991-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...