2,3-Dichloro-5-methylquinoxaline 97%

Supplier: Ambeed
Warning

A320010-100MG A320010-250MG
77263-388EA 50.47 USD
77263-388 77263-390
2,3-Dichloro-5-methylquinoxaline 97%
2,3-Dichloro-5-methylquinoxaline

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂
MW: 213.06 g/mol
MDL Number: MFCD27929282
CAS Number: 82463-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR