(2,4-Difluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Difluorophenylacetic acid

A319992-1G A319992-25G A319992-5G A319992-500G A319992-10G
77239-566EA 5.15 USD
77239-566 77239-568 77239-570 77746-615 77746-614
(2,4-Difluorophenyl)acetic acid 98%
(2,4-Difluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
MDL Number: MFCD00009999
CAS Number: 81228-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR