3,3-Diethoxypropiononitrile 98%

Supplier: Ambeed
Warning

A319584-100G A319584-10G A319584-1G A319584-25G A319584-500G A319584-5G
77277-682EA 196 USD
77277-682 77277-684 77277-686 77277-688 77277-690 77277-692
3,3-Diethoxypropiononitrile 98%
3,3-Diethoxypropiononitrile

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD00040558
CAS Number: 2032-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR