2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate 98%

Supplier: Ambeed
Danger

Synonyms: (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-isothiocyanatotetrahydro-2H-pyran-3,4,5-triyl triacetate, GITC

A319581-100MG A319581-1G A319581-250MG A319581-5G
77251-102EA 80 USD
77251-102 77251-104 77251-106 77251-108
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate 98%
2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₉S
MDL Number: MFCD00043085
CAS Number: 14152-97-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR