Bis(4-bromophenyl)diphenylsilane 97%

Supplier: Ambeed
Warning

A319558-1G A319558-250MG A319558-5G
77239-560EA 118.45 USD
77239-560 77239-562 77239-564
Bis(4-bromophenyl)diphenylsilane 97%
Bis(4-bromophenyl)diphenylsilane

Precaución: for Research Use Only

Formula: C₂₄H₁₈Br₂Si
MW: 494.30 g/mol
MDL Number: MFCD29089347
CAS Number: 18733-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR