2-Benzyloxy-2-methylpropan-1-ol 98%

Supplier: Ambeed
Warning

A319079-100MG A319079-1G A319079-250MG
77263-382EA 43.26 USD
77263-382 77263-384 77263-386
2-Benzyloxy-2-methylpropan-1-ol 98%
2-Benzyloxy-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MDL Number: MFCD08459314
CAS Number: 91968-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR