WAY-262611 98%

Supplier: Ambeed
Warning

A318831-10MG A318831-1MG A318831-50MG A318831-5MG
77239-540EA 147.29 USD
77239-540 77239-542 77239-544 77239-546
WAY-262611 98%
WAY-262611

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄
MW: 318.4 g/mol
MDL Number: MFCD20527803
CAS Number: 1123231-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR