2-Isopropoxyphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropoxybenzeneboronic acid, (2-Isopropoxyphenyl)boronic acid

A318341-25G A318341-1G A318341-10G
77263-358EA 191.58 USD
77263-358 77571-898 77748-625
2-Isopropoxyphenylboronic acid 95%
2-Isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD03427050
CAS Number: 138008-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR