GSK2256098 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-((5-chloro-2-((1-isopropyl-3-methyl-1H-pyrazol-5-yl)amino)pyridin-4-yl)amino)-N-methoxybenzamide

A318281-1MG A318281-25MG A318281-2MG A318281-5MG A318281-10MG
77263-350EA 13.39 USD
77263-350 77263-352 77263-354 77263-356 77616-396
GSK2256098 ≥98%
GSK2256098

Precaución: for Research Use Only

Formula: C₂₀H₂₃ClN₆O₂
MW: 414.89 g/mol
MDL Number: MFCD30502650
CAS Number: 1224887-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR