2-(2-Bromopyridin-4-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromopyridine-4-acetic acid

A318178-1G A318178-250MG
77263-346EA 96.82 USD
77263-346 77263-348
2-(2-Bromopyridin-4-yl)acetic acid 95%
2-(2-Bromopyridin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD07367935
CAS Number: 183483-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR