4-(2-Aminoethyl)-1-benzylpiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Benzyl-4-piperidyl)ethylamine

A318167-1G A318167-250MG A318167-25G A318167-5G
77239-532EA 17 USD
77239-532 77239-534 77239-536 77239-538
4-(2-Aminoethyl)-1-benzylpiperidine 98%
4-(2-Aminoethyl)-1-benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂
MW: 218.34 g/mol
MDL Number: MFCD02093423
CAS Number: 86945-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR