Ethyl 2-chloro-1H-benzo[d]imidazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A317858-100MG A317858-1G A317858-50MG
77263-340EA 47.38 USD
77263-340 77263-342 77263-344
Ethyl 2-chloro-1H-benzo[d]imidazole-6-carboxylate 97%
Ethyl 2-chloro-1H-benzo[d]imidazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.64 g/mol
MDL Number: MFCD12827540
CAS Number: 857035-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR