4-Thien-3-ylbenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Thiophen-3-yl)benzoic acid

A317840-100MG A317840-1G A317840-250MG
77290-256EA 48.41 USD
77290-256 77290-258 77290-260
4-Thien-3-ylbenzoic acid 95%
4-Thien-3-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈O₂S
MW: 204.25 g/mol
MDL Number: MFCD03783559
CAS Number: 29886-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR