4-Bromo-9-phenyl-9H-carbazole 99%

Supplier: Ambeed
Warning

A317780-100MG A317780-1G A317780-250MG A317780-5G
77239-522EA 8 USD
77239-522 77239-524 77239-526 77239-528
4-Bromo-9-phenyl-9H-carbazole 99%
4-Bromo-9-phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂BrN
MDL Number: MFCD29067336
CAS Number: 1097884-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR