3-(Benzyloxymethyl)cyclobutanone 95%

Supplier: Ambeed
Warning

Synonyms: 3-((Benzyloxy)methyl)cyclobutanone

A317765-10G A317765-1G A317765-5G A317765-25G
77277-662EA 136.99 USD
77277-662 77277-664 77277-666 77748-622
3-(Benzyloxymethyl)cyclobutanone 95%
3-(Benzyloxymethyl)cyclobutanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
MW: 190.242 g/mol
MDL Number: MFCD09055138
CAS Number: 172324-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR