2-Chloro-8-nitroquinoxaline 95%

Supplier: Ambeed
Warning

A317552-100MG A317552-1G A317552-250MG
77263-334EA 152.44 USD
77263-334 77263-336 77263-338
2-Chloro-8-nitroquinoxaline 95%
2-Chloro-8-nitroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₄ClN₃O₂
MW: 209.58 g/mol
MDL Number: MFCD29038817
CAS Number: 1934422-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR