Acalisib 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
GS-9820, GS9820, (S)-2-(1-((9H-purin-6-yl)amino)ethyl)-6-fluoro-3-phenylquinazolin-4(3H)-one, GS 9820, 6-fluoro-3-phenyl-2-[(1S)-1-(7H-purin-6-ylamino)ethyl]quinazolin-4-one, Acalisib
A317394-10MG
A317394-1MG
A317394-25MG
A317394-50MG
A317394-5MG
77239-512EA
54.59
USD
77239-512
77239-514
77239-516
77239-518
77239-520
Acalisib 98+%
Acalisib
Precaución: for Research Use Only
Formula:
C₂₁H₁₆FN₇O MW: 401.4 g/mol |
MDL Number:
MFCD28411423 CAS Number: 870281-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...