Methyl 6-methyl-1H-indole-3-carboxylate 98%

Supplier: Ambeed
Warning

A317314-100MG A317314-1G A317314-250MG A317314-5G
77263-322EA 12.36 USD
77263-322 77263-324 77263-326 77263-328
Methyl 6-methyl-1H-indole-3-carboxylate 98%
Methyl 6-methyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD06204260
CAS Number: 163083-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR