Chroman-2-ylmethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A317192-1G A317192-5G
77251-076EA 29 USD
77251-076 77251-078
Chroman-2-ylmethanamine hydrochloride 97%
Chroman-2-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO
MW: 199.67 g/mol
MDL Number: MFCD29059865
CAS Number: 149177-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR