3-Methyl-2-cyclohexen-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-2-cyclohexenone, 3-Methylcyclohex-2-enone

A317175-100G A317175-1G A317175-5G A317175-25G
77277-656EA 129.78 USD
77277-656 77277-658 77277-660 77550-886
3-Methyl-2-cyclohexen-1-one 95%
3-Methyl-2-cyclohexen-1-one

Precaución: for Research Use Only

Formula: C₇H₁₀O
MW: 110.16 g/mol
MDL Number: MFCD00001581
CAS Number: 1193-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR