5-Carboxyoxindole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Oxoindoline-5-carboxylic Acid, 2-Oxindole-5-carboxylic acid

A317039-100MG A317039-1G A317039-250MG A317039-25G A317039-5G
77263-312EA 8.24 USD
77263-312 77263-314 77263-316 77263-318 77748-620
5-Carboxyoxindole 95%
5-Carboxyoxindole

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1600 g/mol
MDL Number: MFCD03411961
CAS Number: 102359-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR