Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%

Supplier: Ambeed
Warning

A316653-100MG A316653-1G A316653-250MG A316653-5G
77263-304EA 52 USD
77263-304 77263-306 77263-308 77263-310
Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate 98%
Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Precaución: for Research Use Only

Formula: C₄₈H₂₈BF₂₄Ir
MDL Number: MFCD10567389
CAS Number: 666826-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR