4-(Trifluoromethyl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)-1,1'-biphenyl

A316632-1G A316632-250MG A316632-5G
77290-234EA 107.12 USD
77290-234 77290-236 77290-238
4-(Trifluoromethyl)biphenyl 97%
4-(Trifluoromethyl)biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉F₃
MW: 222.21 g/mol
MDL Number: MFCD03412287
CAS Number: 398-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR